Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035399.hg.1 |
TC03001978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035399.hg.1 |
ENST00000470487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035399.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197639.hg.1 |
TC0300013058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |