Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011548.hg.1 |
ENST00000299001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011548.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011548.hg.1 |
TC11000921.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011549.hg.1 |
ENST00000299001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011549.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011549.hg.1 |
TC11000921.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100202222.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202222.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202222.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202222.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202222.hg.1 |
TC1100013082.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100202224.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202224.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202224.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202224.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202224.hg.1 |
TC1100013082.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000299001.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000537419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
ENST00000545603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013082.hg.1 |
NM_152431.2 |
Refseq |