Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
TC09000797.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
ENST00000263598.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010347.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011836.hg.1 |
ENST00000372031.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009165.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
ENST00000263598.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
ENST00000371781.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_001252619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
NM_002297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162420.hg.1 |
TC0900009165.hg.1 |
Clariom D TC |