Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058104.hg.1 |
TC01003800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058104.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058104.hg.1 |
ENST00000366997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058104.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230410.hg.1 |
TC0100017223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |