Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
ENST00000398598.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
ENST00000398600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
ENST00000455164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
ENST00000490220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
NM_001144925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
NM_001178046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
NM_002462.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001896.hg.1 |
TC21000189.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000398598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000398600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000413778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000417963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000419044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000424365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000441677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000455164.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000478268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000484465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000490220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_001144925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_001178046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_002462.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
TC2100007208.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000398598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000398600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000413778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000417963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000419044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000424365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000441677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000455164.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000478268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000484465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
ENST00000490220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_001144925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_001178046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148631.hg.1 |
NM_002462.4 |
Refseq |