Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028658.hg.1 |
TC02001782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028658.hg.1 |
ENST00000294973.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028658.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000406924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
ENST00000431905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012383.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000294973.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000402698.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000404451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
ENST00000406007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
NM_012205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190421.hg.1 |
TC0200012383.hg.1 |
Clariom D TC |