Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009343.hg.1 |
TC09000738.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009343.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009343.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000465949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000467100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000472006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
ENST00000492199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008959.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
ENST00000319725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
ENST00000487406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
NM_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160910.hg.1 |
TC0900008959.hg.1 |
Clariom D TC |