Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
TC06001273.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
ENST00000379433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
ENST00000379446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
ENST00000504387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
NM_001142393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
NM_006403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015252.hg.1 |
NM_182966.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
TC06001273.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
ENST00000379433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
ENST00000379446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
ENST00000504387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
NM_001142393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
NM_006403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015250.hg.1 |
NM_182966.3 |
Refseq |