Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
TC03001533.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
ENST00000307227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
ENST00000493112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
ENST00000541608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027939.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185716.hg.1 |
TC0300011436.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000307227.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000460567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000492795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
ENST00000493112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_001177599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011436.hg.1 |
NM_003848.3 |
Refseq |