Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000306167.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000370396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000424519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000424519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000424519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000424519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
ENST00000490530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008716.hg.1 |
NM_000252.2 |
Refseq |