Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005261797.1 | XP_005261854.1 | 9 - 10 | 976 | 71 | |
XM_005261798.1 | XP_005261855.1 | 10 - 11 | 1092 | 71 | ||
XM_005261799.1 | XP_005261856.1 | 9 - 10 | 1119 | 71 | ||
XM_006724347.1 | XP_006724410.1 | 10 - 11 | 1206 | 71 | ||
XM_017029027.1 | XP_016884516.1 | 10 - 11 | 1578 | 71 | ||
GenBank mRNA | AK122673.1 | - | 10 - 11 | 1586 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |