Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198665.1 | NP_001185594.1 | 31 - 32 | 3391 | 145 | |
NM_001301084.1 | NP_001288013.1 | 32 - 33 | 3274 | 145 | ||
NM_015313.2 | NP_056128.1 | 32 - 33 | 3448 | 145 | ||
XM_006718805.3 | XP_006718868.1 | 31 - 32 | 3732 | 145 | ||
XM_011542720.2 | XP_011541022.1 | 32 - 33 | 3389 | 145 | ||
XM_017017420.1 | XP_016872909.1 | 30 - 31 | 3904 | 145 | ||
XM_017017421.1 | XP_016872910.1 | 32 - 33 | 3664 | 145 | ||
XM_017017422.1 | XP_016872911.1 | 31 - 32 | 3156 | 145 | ||
GenBank mRNA | AB002380.2 | - | 27 - 28 | 2810 | 145 | |
AF180681.1 | - | 32 - 33 | 3120 | 145 | ||
AK294803.1 | - | 32 - 33 | 3249 | 145 | ||
AK304316.1 | - | 32 - 33 | 3672 | 145 | ||
BX647785.1 | - | 32 - 33 | 3250 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Homo sapiens Probe spans exons |