Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018160.hg.1 |
TC02001193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018160.hg.1 |
ENST00000303116.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018160.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018160.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174246.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174246.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174246.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174246.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174246.hg.1 |
TC0200010502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000392238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
ENST00000481658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010502.hg.1 |
NM_173511.3 |
Refseq |