Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026732.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026732.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026732.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026732.hg.1 |
TC11002237.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193302.hg.1 |
TC1100012134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000326227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000532855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
ENST00000571244.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012134.hg.1 |
NM_002426.5 |
Refseq |