Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001585.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001585.hg.1 |
ENST00000462997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001585.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001585.hg.1 |
TC20000123.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
TC2000006829.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146735.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000377603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000461589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000462997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000475192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000489753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
ENST00000497333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_001282526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NM_006466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006829.hg.1 |
NR_104209.1 |
Refseq |