Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012918.hg.1 |
TC06001094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012918.hg.1 |
ENST00000358517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012918.hg.1 |
ENST00000367328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012918.hg.1 |
ENST00000535018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012918.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167372.hg.1 |
TC0600009843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000358517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000367328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
ENST00000475490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009843.hg.1 |
NM_001029884.2 |
Refseq |