Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000340160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000517621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000518429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000519051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000520999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000521990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
ENST00000533786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007400.hg.1 |
NM_130783.4 |
Refseq |