Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101417.3 | NP_001094887.1 | 4 - 5 | 689 | 156 | |
NM_001101426.3 | NP_001094896.1 | 5 - 6 | 839 | 156 | ||
XM_006715770.3 | XP_006715833.1 | 5 - 6 | 1009 | 156 | ||
XM_011515499.2 | XP_011513801.1 | 5 - 6 | 984 | 156 | ||
XM_011515500.2 | XP_011513802.1 | 4 - 5 | 879 | 156 | ||
XM_011515508.2 | XP_011513810.1 | 5 - 6 | 1341 | 156 | ||
XM_011515509.2 | XP_011513811.1 | 5 - 6 | 2190 | 156 | ||
XM_017012575.1 | XP_016868064.1 | 5 - 6 | 984 | 156 | ||
XM_017012577.1 | XP_016868066.1 | 4 - 5 | 1101 | 156 | ||
XM_017012578.1 | XP_016868067.1 | 3 - 4 | 1516 | 156 | ||
GenBank mRNA | DY654823.1 | - | 2 - 3 | 92 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |