Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
TC0X000999.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
ENST00000247161.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
ENST00000376983.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
ENST00000542746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013267.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013270.hg.1 |
TC0X000999.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013270.hg.1 |
ENST00000247161.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013270.hg.1 |
ENST00000376983.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013270.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013270.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000343894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
ENST00000468956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_001257168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009581.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
ENST00000247161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
ENST00000376983.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
NM_001114123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
NM_005229.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165288.hg.1 |
TC0X00009581.hg.1 |
Clariom D TC |