Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099433.1 | NP_001092903.1 | 11 - 12 | 2127 | 86 | |
NM_001306133.1 | NP_001293062.1 | 11 - 12 | 1917 | 86 | ||
NM_001306134.1 | NP_001293063.1 | 10 - 11 | 1422 | 86 | ||
NM_144720.3 | NP_653321.1 | 11 - 12 | 2127 | 86 | ||
XM_011513400.2 | XP_011511702.1 | 10 - 11 | 1596 | 86 | ||
XM_017007788.1 | XP_016863277.1 | 11 - 12 | 2091 | 86 | ||
XM_017007789.1 | XP_016863278.1 | 11 - 12 | 1915 | 86 | ||
XM_017007790.1 | XP_016863279.1 | 11 - 12 | 2067 | 86 | ||
XM_017007791.1 | XP_016863280.1 | 11 - 12 | 2091 | 86 | ||
XM_017007792.1 | XP_016863281.1 | 11 - 12 | 2091 | 86 | ||
XM_017007793.1 | XP_016863282.1 | 11 - 12 | 2091 | 86 | ||
XM_017007794.1 | XP_016863283.1 | 11 - 12 | 2091 | 86 | ||
XM_017007795.1 | XP_016863284.1 | 11 - 12 | 2091 | 86 | ||
XM_017007796.1 | XP_016863285.1 | 11 - 12 | 2091 | 86 | ||
XM_017007797.1 | XP_016863286.1 | 10 - 11 | 1596 | 86 | ||
XM_017007798.1 | XP_016863287.1 | 10 - 11 | 2234 | 86 | ||
XM_017007799.1 | XP_016863288.1 | 10 - 11 | 1596 | 86 | ||
GenBank mRNA | AK056126.1 | - | 11 - 12 | 2091 | 86 | |
AK098536.1 | - | 9 - 10 | 1120 | 86 | ||
AK124718.1 | - | 11 - 12 | 1915 | 86 | ||
AK294356.1 | - | 12 - 13 | 1744 | 86 | ||
AK295339.1 | - | 10 - 11 | 1422 | 86 | ||
AK307983.1 | - | 10 - 11 | 1342 | 86 | ||
AM412307.1 | - | 11 - 12 | 2091 | 86 | ||
AM412310.1 | - | 9 - 10 | 1536 | 86 | ||
AW157571.1 | - | 2 - 3 | 86 | |||
AY382340.1 | - | 10 - 11 | 1641 | 86 | ||
BC047075.1 | - | 11 - 12 | 1847 | 5314069 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |