Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010752.hg.1 |
TC01000663.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010752.hg.1 |
ENST00000329505.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010752.hg.1 |
ENST00000336230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010752.hg.1 |
ENST00000360840.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010752.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159676.hg.1 |
TC0100008383.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000336230.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000360840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000398219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000487096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000490670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
ENST00000605337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008383.hg.1 |
NM_016491.3 |
Refseq |