Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012554.hg.1 |
ENST00000267197.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012554.hg.1 |
ENST00000542440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012554.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012554.hg.1 |
TC12000963.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012555.hg.1 |
ENST00000267197.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012555.hg.1 |
ENST00000542440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012555.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012555.hg.1 |
TC12000963.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000267197.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000542440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000604567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000619791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000267197.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000542440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000604567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
ENST00000619791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167963.hg.1 |
TC1200009183.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000267197.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000542440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000604567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000619791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000267197.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000542440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000604567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
ENST00000619791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
NM_015048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167962.hg.1 |
TC1200009183.hg.1 |
Clariom D TC |