Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
TC02001857.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
ENST00000401408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
ENST00000406076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
ENST00000406437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
ENST00000407122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
ENST00000494539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029638.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
TC02001857.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
ENST00000401408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
ENST00000406076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
ENST00000406437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
ENST00000407122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
ENST00000494539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029637.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000401408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000406076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000407122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000411884.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000428621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000494539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000401408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000406076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000407122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000411884.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000428621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
ENST00000494539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192123.hg.1 |
TC0200012639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000401408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000406076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000407122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000411884.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000428621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000487320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000494539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000401408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000406076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000407122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000411884.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000428621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000487320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
ENST00000494539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
NM_001135598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
NM_152385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192124.hg.1 |
TC0200012639.hg.1 |
Clariom D TC |