Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
TC02001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
ENST00000281382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
ENST00000306465.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029074.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
TC02001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
ENST00000281382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
ENST00000306465.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029076.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000420164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000474980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000482786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012489.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191138.hg.1 |
TC0200012489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000281382.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000306465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000412717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
ENST00000495933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
NM_002643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
NM_173074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191140.hg.1 |
TC0200012489.hg.1 |
Clariom D TC |