Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321378.1 | NP_001308307.1 | 11 - 12 | 2178 | 133 | |
NM_001321380.1 | NP_001308309.1 | 9 - 10 | 979 | 133 | ||
NM_002645.3 | NP_002636.2 | 10 - 11 | 2109 | 133 | ||
XM_011520186.2 | XP_011518488.1 | 10 - 11 | 2104 | 133 | ||
XM_017017924.1 | XP_016873413.1 | 11 - 12 | 2125 | 133 | ||
GenBank mRNA | AK294416.1 | - | 9 - 10 | 966 | 133 | |
BC113658.1 | - | 9 - 10 | 1961 | 8322710 | 133 | |
Y13367.1 | - | 9 - 10 | 1895 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |