Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK225967.1 | - | 2 - 3 | 213 | 59 | |
BP213901.1 | - | 2 - 3 | 232 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |