Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024518.2 | NP_078794.1 | 3 - 4 | 709 | 74 | |
GenBank mRNA | AB052908.1 | - | 3 - 4 | 628 | 74 | |
AF304379.1 | - | 3 - 4 | 628 | 74 | ||
AF346596.1 | - | 3 - 4 | 604 | 74 | ||
AK302076.1 | - | 4 - 5 | 595 | 74 | ||
AK315275.1 | - | 5 - 6 | 949 | 74 | ||
AY027538.1 | - | 3 - 4 | 598 | 74 | ||
BC098127.1 | - | 3 - 4 | 628 | 40003400 | 74 | |
CA488356.1 | - | 2 - 3 | 362 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |