Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012061.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012061.hg.1 |
ENST00000356562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012061.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012061.hg.1 |
TC20000886.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012064.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012064.hg.1 |
ENST00000356562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012064.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012064.hg.1 |
TC20000886.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012062.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012062.hg.1 |
ENST00000356562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012062.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012062.hg.1 |
TC20000886.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
TC2000009238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164604.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
TC2000009238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164606.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
TC2000009238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164609.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
TC2000009238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164605.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164610.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164610.hg.1 |
TC2000009238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164610.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000324384.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000356562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
ENST00000618797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009238.hg.1 |
NM_147197.2 |
Refseq |