Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005981.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005981.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005981.hg.1 |
TC16000433.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007784.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156702.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156702.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156702.hg.1 |
ENST00000299191.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156702.hg.1 |
NM_144602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156702.hg.1 |
TC1600007784.hg.1 |
Clariom D TC |