Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011999.hg.1 |
TC04000874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011999.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011999.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400164448.hg.1 |
TC0400009396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000264596.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
ENST00000513321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009396.hg.1 |
NM_018248.2 |
Refseq |