Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023637.hg.1 |
ENST00000310325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023637.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023637.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023637.hg.1 |
TC11001971.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186960.hg.1 |
TC1100011294.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000310325.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000524994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000525733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000526010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000527141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000529561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000530565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
ENST00000533168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011294.hg.1 |
NM_003793.3 |
Refseq |