Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006780.hg.1 |
TC08000497.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006780.hg.1 |
ENST00000260113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006780.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006780.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007993.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
ENST00000260113.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
ENST00000523773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
NM_015886.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800155029.hg.1 |
TC0800007993.hg.1 |
Clariom D TC |