Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010713.hg.1 |
ENST00000491826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012377.hg.1 |
ENST00000399891.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012377.hg.1 |
ENST00000399891.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016473.hg.1 |
TC04001200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016473.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172928.hg.1 |
ENST00000491826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172928.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172928.hg.1 |
ENST00000491826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172928.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172928.hg.1 |
TC0400010713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012377.hg.1 |
ENST00000399891.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012377.hg.1 |
ENST00000399891.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010713.hg.1 |
ENST00000491826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010713.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172929.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172929.hg.1 |
NR_027153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172929.hg.1 |
TC0400010713.hg.1 |
Clariom D TC |