Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023573.hg.1 |
ENST00000259008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023573.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023573.hg.1 |
TC17001769.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700191400.hg.1 |
TC1700011364.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000259008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000577913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000579028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000583837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
ENST00000584322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011364.hg.1 |
NM_032043.2 |
Refseq |