Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
ENST00000326431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007269.hg.1 |
NM_147196.2 |
Refseq |