Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020236.hg.1 |
TC03001169.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020236.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020236.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175069.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175069.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175069.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175069.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175069.hg.1 |
TC0300010282.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
ENST00000287814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010282.hg.1 |
NM_003256.3 |
Refseq |