Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060125.hg.1 |
TC01003974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060125.hg.1 |
ENST00000262861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060125.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060125.hg.1 |
ENST00000366630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060125.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060126.hg.1 |
TC01003974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060126.hg.1 |
ENST00000262861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060126.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060126.hg.1 |
ENST00000366630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060126.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000262861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000366630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000495863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000262861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000366630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
ENST00000495863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233857.hg.1 |
TC0100017748.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000262861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000366630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000262861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000308942.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
ENST00000366630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
NM_020808.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233856.hg.1 |
TC0100017748.hg.1 |
Clariom D TC |