Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293083.1 | NP_001280012.1 | 47 - 48 | 5358 | 70 | |
XM_011512110.2 | XP_011510412.1 | 47 - 48 | 5402 | 70 | ||
XM_011512111.2 | XP_011510413.1 | 47 - 48 | 5399 | 70 | ||
XM_011512113.2 | XP_011510415.1 | 47 - 48 | 5396 | 70 | ||
XM_011512114.2 | XP_011510416.1 | 47 - 48 | 5372 | 70 | ||
XM_011512115.2 | XP_011510417.1 | 46 - 47 | 5333 | 70 | ||
XM_011512116.2 | XP_011510418.1 | 47 - 48 | 5315 | 70 | ||
XM_011512117.2 | XP_011510419.1 | 46 - 47 | 5297 | 70 | ||
XM_011512118.2 | XP_011510420.1 | 46 - 47 | 5282 | 70 | ||
XM_011512119.2 | XP_011510421.1 | 46 - 47 | 5240 | 70 | ||
XM_011512120.2 | XP_011510422.1 | 46 - 47 | 5293 | 70 | ||
XM_011512122.2 | XP_011510424.1 | 47 - 48 | 5403 | 70 | ||
XM_011512124.2 | XP_011510426.1 | 31 - 32 | 4986 | 70 | ||
XM_017005222.1 | XP_016860711.1 | 47 - 48 | 5399 | 70 | ||
XM_017005223.1 | XP_016860712.1 | 30 - 31 | 4050 | 70 | ||
XM_017005224.1 | XP_016860713.1 | 21 - 22 | 2842 | 70 | ||
GenBank mRNA | AL133022.1 | - | 29 - 30 | 4761 | 70 | |
AY461813.1 | - | 46 - 47 | 5384 | 70 | ||
BC117324.1 | - | 13 - 14 | 1566 | 70 | ||
BC126368.1 | - | 13 - 14 | 1569 | 70 | ||
BC143825.1 | - | 13 - 14 | 1566 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |