Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004687.4 | NP_004678.3 | 14 - 15 | 1769 | 96 | |
XM_005257784.2 | XP_005257841.1 | 13 - 14 | 1918 | 96 | ||
XM_005257785.4 | XP_005257842.1 | 15 - 16 | 1886 | 96 | ||
XM_005257786.4 | XP_005257843.1 | 14 - 15 | 2673 | 96 | ||
XM_006722168.3 | XP_006722231.1 | 14 - 15 | 1713 | 96 | ||
XM_011525460.2 | XP_011523762.1 | 15 - 16 | 2766 | 96 | ||
GenBank mRNA | AB014547.2 | - | 12 - 13 | 1659 | 96 | |
AF264717.1 | - | 14 - 15 | 1712 | 96 | ||
BC035609.1 | - | 14 - 15 | 1730 | 5201989 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |