Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027501.hg.1 |
TC05001974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027501.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027501.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027501.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187759.hg.1 |
TC0500012568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000285873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000522075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
ENST00000523355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_001252156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012568.hg.1 |
NM_015465.4 |
Refseq |