Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164326.1 | NP_001157798.1 | 19 - 20 | 2597 | 101 | |
NM_001300966.1 | NP_001287895.1 | 20 - 21 | 2685 | 101 | ||
XM_005271860.3 | XP_005271917.1 | 20 - 21 | 2798 | 101 | ||
XM_017008971.1 | XP_016864460.1 | 18 - 19 | 2636 | 101 | ||
XM_017008973.1 | XP_016864462.1 | 19 - 20 | 2579 | 101 | ||
GenBank mRNA | X87613.1 | - | 19 - 20 | 2615 | 101 | |
X94234.1 | - | 18 - 19 | 2527 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons |