Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000998.hg.1 |
ENST00000251535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000998.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000998.hg.1 |
TC17000081.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700146389.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146389.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146389.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146389.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146389.hg.1 |
TC1700006717.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000251535.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000406228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
ENST00000480801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006717.hg.1 |
NM_000697.2 |
Refseq |