Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
ENST00000160827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
ENST00000400751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
NM_001256269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
NM_001256270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
NM_007317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004657.hg.1 |
TC16000327.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000160827.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000400751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000561482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000565736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000568312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000569382.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_001256269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_001256270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_007317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000160827.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000400751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000561482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000565736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000568312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
ENST00000569382.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_001256269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_001256270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
NM_007317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153671.hg.1 |
TC1600007425.hg.1 |
Clariom D TC |