Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013242.hg.1 |
ENST00000301364.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013242.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013242.hg.1 |
TC17000991.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013243.hg.1 |
ENST00000301364.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013243.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013243.hg.1 |
TC17000991.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000571806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
ENST00000576202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009430.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172177.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172177.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172177.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172177.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172177.hg.1 |
TC1700009430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
ENST00000301364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
ENST00000575049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
NM_018128.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172178.hg.1 |
TC1700009430.hg.1 |
Clariom D TC |