Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
ENST00000356404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
ENST00000436757.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
ENST00000534749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
NM_001130848.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
NM_004910.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023872.hg.1 |
TC11001986.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000356404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000436757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000526602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000527370.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000534749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
NM_001130848.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
NM_004910.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000356404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000436757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000526602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000527370.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
ENST00000534749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
NM_001130848.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
NM_004910.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187406.hg.1 |
TC1100011337.hg.1 |
Clariom D TC |