Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031935.2 | NP_114141.2 | 30 - 31 | 4864 | 137 | |
XM_011510038.2 | XP_011508340.1 | 30 - 31 | 4881 | 137 | ||
XM_011510041.2 | XP_011508343.1 | 30 - 31 | 4889 | 137 | ||
XM_017002437.1 | XP_016857926.1 | 19 - 20 | 2814 | 137 | ||
GenBank mRNA | AK056557.1 | - | 13 - 14 | 1963 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |