Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056645.hg.1 |
TC01006280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056645.hg.1 |
ENST00000367313.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056645.hg.1 |
ENST00000391967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056645.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244426.hg.1 |
TC0100018550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244423.hg.1 |
TC0100018550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000475136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000488842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
ENST00000633953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018550.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
ENST00000632743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244425.hg.1 |
TC0100018550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000367313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000391967.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
ENST00000631576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
NM_005558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244424.hg.1 |
TC0100018550.hg.1 |
Clariom D TC |