Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | P2RX5-TAX1BP3 readthrough (NMD candidate) | P2RX5-TAX1BP3 | Sense | Chr. 17 - 3662893 - 3696404 on NCBI Build GRCh38 | ||
Downstream | integrin subunit alpha E | ITGAE |
CD103, HUMINAE |
36.9kb | Sense | Chr. 17 - 3714625 - 3803766 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |