Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001057.hg.1 |
ENST00000308027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001057.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001057.hg.1 |
TC17000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001055.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001055.hg.1 |
TC17000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001056.hg.1 |
ENST00000308027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001056.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001056.hg.1 |
TC17000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146497.hg.1 |
TC1700006721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006721.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146494.hg.1 |
TC1700006721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
ENST00000308027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
ENST00000575844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
NM_201566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146495.hg.1 |
TC1700006721.hg.1 |
Clariom D TC |