Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010338.hg.1 |
ENST00000369201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010338.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010338.hg.1 |
TC10000853.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161844.hg.1 |
TC1000009025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000442245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000546446.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000553456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
ENST00000616794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_001165924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009025.hg.1 |
NM_004098.3 |
Refseq |